Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317002.1 | NP_001303931.1 | 4, 5 | 289 | |
NM_177964.4 | NP_808879.2 | 3, 4 | 471 | ||
XM_017003364.1 | XP_016858853.1 | 4, 5 | 299 | ||
GenBank mRNA | BC040176.2 | - | 3, 4 | 471 | |
DB461489.1 | - | 4, 5 | 282 |