Gene Transcripts

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
Chromosome Location
Chr. 10: 70672518 - 70762441 on Build GRCh38
UniGene ID
Hs.352156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080722.4 NP_542453.2 22 3533
NM_139155.3 NP_631894.2 22 3542
XM_011539300.2 XP_011537602.1 19 3016
XM_011539301.2 XP_011537603.1 19 2539
XM_011539302.2 XP_011537604.1 19 2416
XM_011539309.1 XP_011537611.1 15 2126
GenBank mRNA AF358666.1 - 22 3236
AF366351.1 - 21 3163
AJ345098.1 - 22 3236

View Full Product Details