Gene Transcripts

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
Chromosome Location
Chr. 3: 68731764 - 68932610 on Build GRCh38
UniGene ID
Hs.187873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005527.2 NP_001005527.1 5 737
NM_182522.4 NP_872328.1 5 819
XM_011533371.1 XP_011531673.1 5 819
XM_011533372.1 XP_011531674.1 5 695
GenBank mRNA AK057890.1 - 4 435
AY325117.1 - 5 791
BC031566.1 - 5 651 5195201

View Full Product Details