Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr. 4: 47847233 - 47914667 on Build GRCh38
UniGene ID
Hs.646325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278623.1 NP_001265552.1 13 1633
NM_001278624.1 NP_001265553.1 13 1771
NM_152995.5 NP_694540.3 13 1673
NR_103795.1 13 1633

View Full Product Details