Gene Transcripts

Gene Symbol
SPRR4
Entrez Gene ID
163778
Gene Name
small proline rich protein 4
Gene Aliases
Chromosome Location
Chr. 1: 152968765 - 152972602 on Build GRCh38
UniGene ID
Hs.433629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173080.2 NP_775103.1 2 175
XM_017000482.1 XP_016855971.1 3 871
GenBank mRNA BC069445.1 - 2 163
BC069549.1 - 2 163 7262739
BC112170.1 - 2 175 8327638
BC112172.1 - 2 173 8327640
W77994.1 - 1 108

View Full Product Details