Gene Transcripts

Gene Symbol
SASS6
Entrez Gene ID
163786
Gene Name
SAS-6 centriolar assembly protein
Gene Aliases
MCPH14, SAS-6, SAS6
Chromosome Location
Chr. 1: 100082632 - 100133095 on Build GRCh38
UniGene ID
Hs.591447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304829.1 NP_001291758.1 10 1506
NM_194292.2 NP_919268.1 11 1586
XM_017000486.1 XP_016855975.1 11 1352
GenBank mRNA AK302513.1 - 12 1336
AK307824.1 - 10 1372
AL834265.1 - 7 849
AY359522.1 - 11 1446
BC101026.3 - 10 1299 40007432
BX641109.1 - 10 1497
BX648406.1 - 11 1343

View Full Product Details