Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
Chromosome Location
Chr. 2: 119544392 - 119688610 on Build GRCh38
UniGene ID
Hs.496753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK293665.1 - 12 1306
BC036530.1 - 14 1535 4828213

View Full Product Details