Gene Transcripts

Gene Symbol
LYSMD2
Entrez Gene ID
256586
Gene Name
LysM domain containing 2
Gene Aliases
Chromosome Location
Chr. 15: 51723064 - 51751453 on Build GRCh38
UniGene ID
Hs.603629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143917.1 NP_001137389.1 2 462
NM_153374.2 NP_699205.1 2 1053
XM_017022057.1 XP_016877546.1 2 445
GenBank mRNA AK129759.1 - 2 532
AY125955.1 - 2 1042
BC022075.1 - 2 598 4799858
BC033515.1 - 2 1050 5175226
CR936638.1 - 1 92

View Full Product Details