Gene Transcripts

Gene Symbol
MORN3
Entrez Gene ID
283385
Gene Name
MORN repeat containing 3
Gene Aliases
Chromosome Location
Chr. 12: 121651108 - 121672650 on Build GRCh38
UniGene ID
Hs.434154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173855.4 NP_776254.3 4, 5 829
XM_005253869.4 XP_005253926.1 5, 6 758
XM_011538213.2 XP_011536515.1 5, 6 769
XM_011538214.2 XP_011536516.1 2, 3 610
GenBank mRNA BC057760.1 - 4, 5 829

View Full Product Details