Gene Transcripts

Gene Symbol
TMED8
Entrez Gene ID
283578
Gene Name
transmembrane p24 trafficking protein family member 8
Gene Aliases
FAM15B
Chromosome Location
Chr. 14: 77335021 - 77377384 on Build GRCh38
UniGene ID
Hs.200413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213601.1 NP_998766.1 4 401
XM_017021223.1 XP_016876712.1 4 747
XM_017021224.1 XP_016876713.1 3 300
XM_017021225.1 XP_016876714.1 4 2862
GenBank mRNA AK095650.1 - 4 445
AY598329.1 - 4 401
BC101510.1 - 4 457 8069016

View Full Product Details