Gene Transcripts

Gene Symbol
SLC26A11
Entrez Gene ID
284129
Gene Name
solute carrier family 26 member 11
Gene Aliases
Chromosome Location
Chr. 17: 80220287 - 80253509 on Build GRCh38
UniGene ID
Hs.4866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166347.1 NP_001159819.1 9, 10 1296
NM_001166348.1 NP_001159820.1 8, 9 1226
NM_001166349.1 NP_001159821.1 8, 9 1096
NM_173626.3 NP_775897.3 9, 10 1293
XM_006721833.1 XP_006721896.1 9, 10 1368
XM_011524652.1 XP_011522954.1 7, 8 1129
XM_011524653.1 XP_011522955.1 7, 8 1057
XM_011524654.1 XP_011522956.1 7, 8 942
XM_017024505.1 XP_016879994.1 8, 9 1282
XM_017024506.1 XP_016879995.1 8, 9 1282
XM_017024507.1 XP_016879996.1 9, 10 1342
GenBank mRNA AF331524.1 - 1, 2 70
AK075248.1 - 9, 10 1256
AK315132.1 - 9, 10 1259
AL833468.1 - 8, 9 1190
BC035900.1 - 9, 10 1235 5241643
BC047451.1 - 3, 4 345

View Full Product Details