Gene Transcripts

Gene Symbol
ZFP82
Entrez Gene ID
284406
Gene Name
ZFP82 zinc finger protein
Gene Aliases
ZNF545
Chromosome Location
Chr. 19: 36383102 - 36418674 on Build GRCh38
UniGene ID
Hs.558734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321917.1 NP_001308846.1 5 1237
NM_133466.3 NP_597723.1 5 1234
GenBank mRNA AB075828.1 - 1 2701
AK074942.1 - 5 1216
AK315709.1 - 5 1194
AL834267.1 - 5 1208
BC032577.2 - 5 1201 5492906
GU388429.1 - 5 1204

View Full Product Details