Gene Transcripts

Gene Symbol
RUFY4
Entrez Gene ID
285180
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
ZFYVE31
Chromosome Location
Chr. 2: 218034934 - 218090584 on Build GRCh38
UniGene ID
Hs.570069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198483.3 NP_940885.2 9 1667
NR_034176.1 10 2528
XM_011511014.2 XP_011509316.1 11 2609
XM_017003895.1 XP_016859384.1 10 2744
XM_017003896.1 XP_016859385.1 10 2741
XM_017003897.1 XP_016859386.1 10 2744
GenBank mRNA AK128393.1 - 10 2474
AK131246.1 - 8 1619
BC113712.1 - 3 689 8322764

View Full Product Details