Gene Transcripts

Gene Symbol
FBXW12
Entrez Gene ID
285231
Gene Name
F-box and WD repeat domain containing 12
Gene Aliases
FBW12, FBXO12, FBXO35
Chromosome Location
Chr. 3: 48372219 - 48394830 on Build GRCh38
UniGene ID
Hs.288793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159927.1 NP_001153399.1 7, 8 997
NM_001159929.1 NP_001153401.1 7, 8 1006
NM_207102.2 NP_996985.2 8, 9 1180
XM_017006224.1 XP_016861713.1 7, 8 1193
GenBank mRNA AK097594.1 - 2, 3 298
AY247969.1 - 8, 9 1180
BC101303.1 - 7, 8 1353 40024696
BC101304.1 - 7, 8 941 40024699
BC101305.1 - 7, 8 950 40024702
BC101306.1 - 5, 6 688

View Full Product Details