Gene Transcripts

Gene Symbol
FAM19A2
Entrez Gene ID
338811
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Gene Aliases
TAFA-2, TAFA2
Chromosome Location
Chr. 12: 61708248 - 62192839 on Build GRCh38
UniGene ID
Hs.269745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178539.4 NP_848634.1 3 1768
XM_006719370.2 XP_006719433.1 4 1497
XM_011538272.1 XP_011536574.1 4 1830
XM_011538274.1 XP_011536576.1 4 1221
GenBank mRNA AK123580.1 - 4 376
AL834160.1 - 3 1120
AY325115.1 - 3 341
BC028403.1 - 3 717 4818605
BC040286.1 - 2 160
BC050347.1 - 3 1699
CR749367.1 - 6 676

View Full Product Details