Gene Transcripts

Gene Symbol
CCDC36
Entrez Gene ID
339834
Gene Name
coiled-coil domain containing 36
Gene Aliases
CT74
Chromosome Location
Chr. 3: 49198428 - 49258104 on Build GRCh38
UniGene ID
Hs.631931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135197.1 NP_001128669.1 8 1649
NM_178173.3 NP_835467.2 10 1947
XM_006713124.3 XP_006713187.1 8 1824
XM_011533670.2 XP_011531972.1 7 1658
XM_011533671.2 XP_011531973.1 7 1652
XM_011533672.2 XP_011531974.1 8 2017
XM_011533673.2 XP_011531975.1 8 2049
GenBank mRNA AK303033.1 - 8 1649
BC036202.1 - 10 1947 4836976

View Full Product Details