Gene Transcripts

Gene Symbol
KY
Entrez Gene ID
339855
Gene Name
kyphoscoliosis peptidase
Gene Aliases
Chromosome Location
Chr. 3: 134599923 - 134656053 on Build GRCh38
UniGene ID
Hs.146730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178554.4 NP_848649.3 6 481
XM_005247418.3 XP_005247475.1 5 5449
XM_006713612.3 XP_006713675.1 5 5395
XM_017006287.1 XP_016861776.1 6 5527
XM_017006288.1 XP_016861777.1 6 5473
XM_017006289.1 XP_016861778.1 5 5464
XM_017006290.1 XP_016861779.1 5 5410
GenBank mRNA AK090526.1 - 6 481
AK126993.1 - 5 364
AK293840.1 - 5 414
AK294500.1 - 6 482
BC136349.1 - 6 442
DA238247.1 - 6 481

View Full Product Details