Gene Transcripts

Gene Symbol
VSTM2B
Entrez Gene ID
342865
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
Chromosome Location
Chr. 19: 29525431 - 29564555 on Build GRCh38
UniGene ID
Hs.451618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146339.1 NP_001139811.1 2, 3 265
XM_011526902.2 XP_011525204.1 3, 4 885
XM_011526904.2 XP_011525206.1 2, 3 410

View Full Product Details