Gene Transcripts

Gene Symbol
ATP13A5
Entrez Gene ID
344905
Gene Name
ATPase 13A5
Gene Aliases
Chromosome Location
Chr. 3: 193274789 - 193378824 on Build GRCh38
UniGene ID
Hs.658032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198505.2 NP_940907.2 30 3569
XM_017006304.1 XP_016861793.1 26 2882
XM_017006305.1 XP_016861794.1 17 1985
GenBank mRNA AY358667.1 - 4 378
AY823163.1 - 30 3569

View Full Product Details