Gene Transcripts
- Gene Symbol
- INCA1
- Entrez Gene ID
- 388324
- Gene Name
- inhibitor of CDK, cyclin A1 interacting protein 1
- Gene Aliases
- HSD45
- Chromosome Location
- Chr. 17: 4988130 - 4997933 on Build GRCh38
- UniGene ID
- Hs.658926
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167985.1 |
NP_001161457.1
|
6, 7 |
626 |
|
NM_001167986.1 |
NP_001161458.1
|
7, 8 |
958 |
|
NM_001167987.1 |
NP_001161459.1
|
6, 7 |
671 |
|
NM_213726.2 |
NP_998891.2
|
6, 7 |
788 |
|
XM_005256627.3 |
XP_005256684.1
|
5, 6 |
866 |
|
XM_005256628.4 |
XP_005256685.1
|
5, 6 |
1063 |
|
XM_005256629.1 |
XP_005256686.1
|
5, 6 |
873 |
|
XM_006721517.1 |
XP_006721580.1
|
5, 6 |
918 |
|
XM_006721518.2 |
XP_006721581.1
|
5, 6 |
822 |
|
XM_006721519.3 |
XP_006721582.1
|
5, 6 |
1019 |
|
XM_011523832.2 |
XP_011522134.1
|
6, 7 |
1188 |
|
GenBank mRNA |
AY601906.1 |
- |
6, 7 |
788 |
|
AY601907.1 |
- |
6, 7 |
626 |
|
AY604176.1 |
- |
4, 5 |
346 |
|
BC119781.1 |
- |
6, 7 |
747 |
40109268 |
BC122873.1 |
- |
5, 6 |
460 |
40109270 |
View Full Product Details