Gene Transcripts

Gene Symbol
RFX8
Entrez Gene ID
731220
Gene Name
RFX family member 8, lacking RFX DNA binding domain
Gene Aliases
Chromosome Location
Chr. 2: 101397344 - 101475179 on Build GRCh38
UniGene ID
Hs.662488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145664.1 NP_001139136.1 12 1443
XM_011511771.2 XP_011510073.1 13 1981
XM_011511772.1 XP_011510074.1 15 1655
XM_011511773.1 XP_011510075.1 11 1326
XM_011511776.1 XP_011510078.1 14 1614
XM_011511777.1 XP_011510079.1 9 1984
XM_011511778.1 XP_011510080.1 9 1321
XM_017004851.1 XP_016860340.1 15 1665
XM_017004852.1 XP_016860341.1 12 1452
XM_017004853.1 XP_016860342.1 14 1348
GenBank mRNA AK124976.1 - 8 2498
AK131289.1 - 14 1621
AK298610.1 - 12 1443

View Full Product Details