Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006432.3 | NP_006423.1 | 4 | 499 | |
XM_017020930.1 | XP_016876419.1 | 4 | 593 | ||
GenBank mRNA | AK222474.1 | - | 4 | 419 | |
AK300879.1 | - | 4 | 426 | ||
AK311931.1 | - | 4 | 460 | ||
BC002532.2 | - | 4 | 498 | 3140870 | |
X67698.1 | - | 4 | 402 |