Gene Transcripts
- Gene Symbol
- ZNF818P
- Entrez Gene ID
- 390963
- Gene Name
- zinc finger protein 818, pseudogene
- Gene Aliases
- ZNF818
- Chromosome Location
- Chr. 19: 53212948 - 53216102 on Build GRCh38
- UniGene ID
- Hs.444446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073396.1 |
|
1 |
246 |
|
GenBank mRNA |
AK128250.1 |
- |
1 |
219 |
|
BF908632.1 |
- |
1 |
262 |
|
View Full Product Details