Gene Transcripts

Gene Symbol
TARDBP
Entrez Gene ID
23435
Gene Name
TAR DNA binding protein
Gene Aliases
ALS10, TDP-43
Chromosome Location
Chr. 1: 11012622 - 11030528 on Build GRCh38
UniGene ID
Hs.300624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007375.3 NP_031401.1 6 940
XM_017000863.1 XP_016856352.1 6 928
XM_017000864.1 XP_016856353.1 6 928
XM_017000865.1 XP_016856354.1 6 928
XM_017000866.1 XP_016856355.1 6 928
XM_017000867.1 XP_016856356.1 6 928
XM_017000868.1 XP_016856357.1 6 928
GenBank mRNA AK222754.1 - 6 878
AK295920.1 - 5 714
AK299454.1 - 6 691
AK312416.1 - 6 908
AL050265.1 - 6 894
BC001487.2 - 6 893
BC071657.1 - 6 891 5498250
BC095435.1 - 6 892 30389805
CR533534.1 - 5 806
EF434181.1 - 5 806
EF434182.1 - 5 806
EF434183.1 - 4 449
HQ628633.1 - 4 642
HQ628634.1 - 6 715
HQ628635.1 - 5 806
U23731.1 - 5 806

View Full Product Details