Gene Transcripts
- Gene Symbol
- GSTZ1
- Entrez Gene ID
- 2954
- Gene Name
- glutathione S-transferase zeta 1
- Gene Aliases
- GSTZ1-1, MAAI, MAI
- Chromosome Location
- Chr. 14: 77320887 - 77331597 on Build GRCh38
- UniGene ID
- Hs.655292
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001312660.1 |
NP_001299589.1
|
2, 3 |
491 |
|
NM_145870.2 |
NP_665877.1
|
1, 2 |
305 |
|
NM_145871.2 |
NP_665878.2
|
1, 2 |
305 |
|
XM_011536670.2 |
XP_011534972.1
|
2, 3 |
547 |
|
XM_011536671.2 |
XP_011534973.1
|
1, 2 |
215 |
|
XM_017021243.1 |
XP_016876732.1
|
1, 2 |
217 |
|
GenBank mRNA |
AJ001838.1 |
- |
1, 2 |
126 |
|
AK309054.1 |
- |
1, 2 |
153 |
|
AK315154.1 |
- |
2, 3 |
382 |
|
AK315609.1 |
- |
1, 2 |
149 |
|
BC001453.1 |
- |
1, 2 |
181 |
3139094 |
BE958109.1 |
- |
|
112 |
|
BG330365.1 |
- |
1, 2 |
167 |
|
BG772450.1 |
- |
1, 2 |
312 |
|
BI559916.1 |
- |
2, 3 |
400 |
|
BI829992.1 |
- |
2, 3 |
371 |
|
CR456987.1 |
- |
|
23 |
|
U86529.1 |
- |
1, 2 |
126 |
|
View Full Product Details