Gene Transcripts

Gene Symbol
RAD51
Entrez Gene ID
5888
Gene Name
RAD51 recombinase
Gene Aliases
BRCC5, FANCR, HRAD51, HsRad51, HsT16930, MRMV2, RAD51A, RECA
Chromosome Location
Chr. 15: 40694774 - 40732340 on Build GRCh38
UniGene ID
Hs.631709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164269.1 NP_001157741.1 5 502
NM_001164270.1 NP_001157742.1 5 654
NM_002875.4 NP_002866.2 5 654
NM_133487.3 NP_597994.3 5 657
XM_006720626.3 XP_006720689.1 5 705
XM_011521857.2 XP_011520159.1 5 404
XM_011521858.2 XP_011520160.1 5 451
XM_011521859.2 XP_011520161.1 5 588
XM_011521860.2 XP_011520162.1 5 499
XM_011521861.2 XP_011520163.1 5 404
XM_011521862.2 XP_011520164.1 4 504
GenBank mRNA AK131299.1 - 5 502
AK291969.1 - 5 612
AK313503.1 - 5 609
AY425955.1 - 4 322
AY608888.1 - 2 69
BT019705.1 - 4 355
CR536559.1 - 4 355
D13804.1 - 5 566
D14134.1 - 5 587
EU362635.1 - 5 654

View Full Product Details