Gene Transcripts

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
Chromosome Location
Chr. 17: 8224821 - 8248095 on Build GRCh38
UniGene ID
Hs.156055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025099.5 NP_079375.3 6 920
NR_046431.1 6 815
XM_006721577.3 XP_006721640.1 6 904
XM_006721578.3 XP_006721641.1 6 904
XM_011524010.2 XP_011522312.1 6 799
XM_011524011.2 XP_011522313.1 5 669
GenBank mRNA AK091077.1 - 6 871
AL831955.1 - 6 762
BC111783.1 - 6 871 40054070
DB126489.1 - 2 119
DQ451688.1 - 6 861
FJ790670.1 - 6 861

View Full Product Details