Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012727.1 | NP_001012745.1 | 5 | 1100 | |
NM_006412.3 | NP_006403.2 | 6 | 1196 | ||
GenBank mRNA | AF000237.1 | - | 6 | 1160 | |
AF011374.1 | - | 6 | 1157 | ||
BC000026.2 | - | 6 | 1139 | 3507150 | |
BC004529.2 | - | 5 | 1034 | 3347049 | |
BC007269.1 | - | 4 | 771 | ||
BC019292.1 | - | 6 | 1154 | 2823580 | |
U56418.2 | - | 6 | 1133 |