Gene Transcripts

Gene Symbol
FOLR3
Entrez Gene ID
2352
Gene Name
folate receptor 3
Gene Aliases
FR-G, FR-gamma, gamma-hFR
Chromosome Location
Chr. 11: 72135710 - 72139892 on Build GRCh38
UniGene ID
Hs.352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000804.3 NP_000795.2 5 667
NM_001318045.1 NP_001304974.1 5 795
XM_011544876.1 XP_011543178.1 3 331
GenBank mRNA BC030285.1 - 5 793
BI836723.1 - 5 792
U08470.1 - 4 611
U08471.1 - 4 613
Z32564.1 - 4 613
Z32633.1 - 4 611

View Full Product Details