Gene Transcripts

Gene Symbol
CD3D
Entrez Gene ID
915
Gene Name
CD3d molecule
Gene Aliases
CD3-DELTA, IMD19, T3D
Chromosome Location
Chr. 11: 118339074 - 118344233 on Build GRCh38
UniGene ID
Hs.504048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000732.4 NP_000723.1 2 293
NM_001040651.1 NP_001035741.1 2 293
XM_017018543.1 XP_016874032.1 3 1193
GenBank mRNA AK311971.1 - 2 186
BC039035.1 - 2 157 5745355
BC070321.1 - 2 293 30412345
CD014058.1 - 2 180

View Full Product Details