Gene Transcripts

Gene Symbol
RAB23
Entrez Gene ID
51715
Gene Name
RAB23, member RAS oncogene family
Gene Aliases
HSPC137
Chromosome Location
Chr. 6: 57186992 - 57222314 on Build GRCh38
UniGene ID
Hs.555016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278666.1 NP_001265595.1 2 156
NM_001278667.1 NP_001265596.1 2 328
NM_001278668.1 NP_001265597.1 2 155
NM_016277.4 NP_057361.3 2 625
NM_183227.2 NP_899050.1 2 179
NR_103822.1 2 156
GenBank mRNA AB034244.1 - 2 125
AF161486.1 - 2 121
AK311123.1 - 2 145
AK313796.1 - 2 176
AU280196.1 - 2 155
BC015021.1 - 2 591 3910708
CR749371.1 - 3 359
DA021294.1 - 2 179
DA997359.1 - 2 324

View Full Product Details