Gene Transcripts

Gene Symbol
FUT3
Entrez Gene ID
2525
Gene Name
fucosyltransferase 3 (Lewis blood group)
Gene Aliases
CD174, FT3B, FucT-III, LE, Les
Chromosome Location
Chr. 19: 5842888 - 5851474 on Build GRCh38
UniGene ID
Hs.169238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000149.3 NP_000140.1 3 1814
NM_001097639.1 NP_001091108.1 3 1466
NM_001097640.1 NP_001091109.1 3 1412
NM_001097641.1 NP_001091110.1 2 1273
XM_011527865.1 XP_011526167.1 3 1792
XM_011527866.1 XP_011526168.1 3 1453
XM_011527867.1 XP_011526169.1 3 1422
GenBank mRNA AF131913.1 - 2 1345
AK291852.1 - 3 1441
BC108675.1 - 2 1270 5187745
U27326.1 - 3 1405
U27327.1 - 3 1459
U27328.1 - 4 1808
X53578.1 - 2 1248

View Full Product Details