Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr. 22: 24719034 - 24774720 on Build GRCh38
UniGene ID
Hs.448343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008496.3 NP_001008496.2 19, 20 2799
NM_001255975.1 NP_001242904.1 19, 20 2772
NR_045648.1 20, 21 2982
NR_045649.1 20, 21 2855
GenBank mRNA AB079368.1 - 19, 20 2795
AK302409.1 - 19, 20 2772
AK315806.1 - 19, 20 2772
BC144693.1 - 20, 21 2612
BC144696.1 - 20, 21 2739

View Full Product Details