Gene Transcripts

Gene Symbol
MBOAT4
Entrez Gene ID
619373
Gene Name
membrane bound O-acyltransferase domain containing 4
Gene Aliases
FKSG89, GOAT, OACT4
Chromosome Location
Chr. 8: 30131454 - 30148768 on Build GRCh38
UniGene ID
Hs.627194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100916.1 NP_001094386.1 3 1313
XM_017013726.1 XP_016869215.1 4 1916
GenBank mRNA AF359269.1 - 1 933
EU518495.2 - 3 1323

View Full Product Details