Gene Transcripts

Gene Symbol
GPR89A
Entrez Gene ID
653519
Gene Name
G protein-coupled receptor 89A
Gene Aliases
GPHR, GPR89, GPR89B, SH120, UNQ192
Chromosome Location
Chr. 1: 145607960 - 145670977 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097612.1 NP_001091081.1 6, 7 719
NM_001097613.2 NP_001091082.2 7, 8 765
NR_036541.1 6, 7 739
XM_006711492.3 XP_006711555.1 6, 7 689
XM_006711493.3 XP_006711556.1 7, 8 697
XM_011509908.2 XP_011508210.1 7, 8 735
XM_011509909.2 XP_011508211.1 6, 7 947
XM_011509910.2 XP_011508212.1 6, 7 689
XM_017002150.1 XP_016857639.1 6, 7 689
GenBank mRNA AB097024.1 - 6, 7 660
AB097025.1 - 6, 7 660
AF132947.1 - 6, 7 644
AK021758.1 - 6, 7 709
AK223223.1 - 6, 7 660
AK297145.1 - 6, 7 739
AK301939.1 - 6, 7 566
AK316371.1 - 7, 8 677
AY358966.1 - 6, 7 694
BC003187.1 - 6, 7 622 3537949
BC067816.1 - 6, 7 678 5295014
BC146880.1 - 6, 7 640 9056808
BT006679.1 - 6, 7 545
KJ534847.1 - 6, 7 545
KJ534848.1 - 6, 7 545
KJ535029.1 - 6, 7 545
KJ535049.1 - 6, 7 565

Gene Symbol
GPR89B
Entrez Gene ID
51463
Gene Name
G protein-coupled receptor 89B
Gene Aliases
GPHR, GPR89, GPR89C, SH120, UNQ192
Chromosome Location
Chr. 1: 147928393 - 147993523 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016334.3 NP_057418.1 6, 7 689
XM_005277401.3 XP_005277458.1 6, 7 662
XM_005277402.4 XP_005277459.1 6, 7 826
XM_005277404.3 XP_005277461.1 6, 7 682
XM_006711377.3 XP_006711440.1 7, 8 725
XM_006711378.2 XP_006711441.1 7, 8 728
XM_011509610.2 XP_011507912.1 7, 8 722
XM_011509612.2 XP_011507914.1 7, 8 683
XM_011509613.2 XP_011507915.1 6, 7 662
XM_017001448.1 XP_016856937.1 7, 8 1225
XM_017001449.1 XP_016856938.1 6, 7 662
GenBank mRNA AK299995.1 - 6, 7 660
AK302266.1 - 6, 7 681
AL133020.1 - 6, 7 673
CD102528.1 - 1, 2 54
KJ534849.1 - 6, 7 545
KJ534850.1 - 6, 7 545
KJ534851.1 - 6, 7 476

View Full Product Details