Gene Transcripts

Gene Symbol
TARP
Entrez Gene ID
445347
Gene Name
TCR gamma alternate reading frame protein
Gene Aliases
CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
Chromosome Location
Chr. 7: 38239577 - 38276741 on Build GRCh38
UniGene ID
Hs.534032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA S60175.1 - 1 87
S73842.1 - 1 87
Y00790.1 - 2 288

Gene Symbol
TRGC2
Entrez Gene ID
6967
Gene Name
T cell receptor gamma constant 2
Gene Aliases
TCRGC2, TRGC2(2X), TRGC2(3X)
Chromosome Location
Chr. 7: 38240024 - 38249572 on Build GRCh38
UniGene ID
Hs.731375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M13231.1 - 2 255

Gene Symbol
TRGV3
Entrez Gene ID
6976
Gene Name
T cell receptor gamma variable 3
Gene Aliases
TCRGV3, V1S3
Chromosome Location
Chr. 7: 38358512 - 38358979 on Build GRCh38
UniGene ID
Hs.731375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC071739.1 - 2 435

Gene Symbol
TRGV5
Entrez Gene ID
6978
Gene Name
T cell receptor gamma variable 5
Gene Aliases
TCRGV5, V1S5
Chromosome Location
Chr. 7: 38349355 - 38349824 on Build GRCh38
UniGene ID
Hs.731375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC035592.1 - 2 441

View Full Product Details