Gene Transcripts

Gene Symbol
TRAK1
Entrez Gene ID
22906
Gene Name
trafficking kinesin protein 1
Gene Aliases
MILT1, OIP106
Chromosome Location
Chr. 3: 42013802 - 42225890 on Build GRCh38
UniGene ID
Hs.535711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017005906.1 XP_016861395.1 1, 2 315
XM_017005909.1 XP_016861398.1 1, 2 315
XM_017005911.1 XP_016861400.1 1, 2 315

View Full Product Details