Gene Transcripts
- Gene Symbol
- HYDIN2
- Entrez Gene ID
- 100288805
- Gene Name
- HYDIN2, axonemal central pair apparatus protein (pseudogene)
- Gene Aliases
- HYDINP1
- Chromosome Location
- Chr. 1: 146547489 - 146822034 on Build GRCh38
- UniGene ID
- Hs.648712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103556.1 |
|
11 |
1532 |
|
GenBank mRNA |
AL833826.1 |
- |
13 |
1728 |
|
AL834340.1 |
- |
8 |
1022 |
|
View Full Product Details