Gene Transcripts
- Gene Symbol
- LOC100996274
- Entrez Gene ID
- 100996274
- Gene Name
- uncharacterized LOC100996274
- Gene Aliases
- Chromosome Location
- Chr. 18: 79144752 - 79149853 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- ATP9B
- Entrez Gene ID
- 374868
- Gene Name
- ATPase phospholipid transporting 9B (putative)
- Gene Aliases
- ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
- Chromosome Location
- Chr. 18: 79069275 - 79378391 on Build GRCh38
- UniGene ID
- Hs.465475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128847.1 |
- |
1 |
2067 |
|
View Full Product Details