Gene Transcripts

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
Chromosome Location
Chr. 9: 128504692 - 128542301 on Build GRCh38
UniGene ID
Hs.522418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003722.1 NP_001003722.1 1, 2 218
NM_001499.2 NP_001490.1 1, 2 218
XM_011518551.1 XP_011516853.1 1, 2 218
GenBank mRNA AF058922.1 - 1, 2 191
AK222842.1 - 1, 2 164
AK222854.1 - 1, 2 192
AK290533.1 - 1, 2 104
BC030012.1 - 1, 2 211 4829102
BM786801.1 - 1, 2 218

View Full Product Details