Gene Transcripts

Gene Symbol
KLHL2
Entrez Gene ID
11275
Gene Name
kelch like family member 2
Gene Aliases
ABP-KELCH, MAV, MAYVEN
Chromosome Location
Chr. 4: 165207618 - 165323156 on Build GRCh38
UniGene ID
Hs.388668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161521.1 NP_001154993.1 5 649
NM_001161522.1 NP_001154994.1 4 542
NM_007246.3 NP_009177.3 5 772
XM_011531572.2 XP_011529874.1 6 762
XM_011531575.2 XP_011529877.1 4 594
XM_011531576.2 XP_011529878.1 3 262
XM_017007674.1 XP_016863163.1 6 722
XM_017007675.1 XP_016863164.1 4 487
XM_017007676.1 XP_016863165.1 4 595
XM_017007677.1 XP_016863166.1 3 469
GenBank mRNA AB209727.1 - 1 116
AF059569.1 - 5 621
AK025289.1 - 2 238
AK294103.1 - 5 643
AK294336.1 - 1 71
AK308940.1 - 1 72
AK315372.1 - 5 617
AL541403.3 - 5 772
BC036468.2 - 5 713 4799622
BX647776.1 - 4 526

View Full Product Details