Gene Transcripts

Gene Symbol
LOC102724467
Entrez Gene ID
102724467
Gene Name
uncharacterized LOC102724467
Gene Aliases
Chromosome Location
Chr. 16: 87766331 - 87779148 on Build GRCh38
UniGene ID
Hs.587857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120309.1 2 404
GenBank mRNA DB094384.1 - 2 406
DB100335.1 - 2 451
HY133504.1 - 1 117

View Full Product Details