Gene Transcripts

Gene Symbol
MAEA
Entrez Gene ID
10296
Gene Name
macrophage erythroblast attacher
Gene Aliases
EMLP, EMP, GID9, HLC-10, PIG5
Chromosome Location
Chr. 4: 1289851 - 1340148 on Build GRCh38
UniGene ID
Hs.139896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017405.2 NP_001017405.1 9 1907
NM_001297430.1 NP_001284359.1 8 1703
NM_001297431.1 NP_001284360.1 7 1703
NM_001297432.1 NP_001284361.1 9 2045
NM_001297433.1 NP_001284362.1 9 1981
NM_005882.4 NP_005873.2 8 1784
NR_123716.1 7 1707
XM_017007636.1 XP_016863125.1 10 2027
XM_017007637.1 XP_016863126.1 9 1944
GenBank mRNA AB208907.1 - 3 5063
AK001088.1 - 9 1867
AK022515.1 - 7 1672
AK022586.1 - 8 1744
AK091951.1 - 4 1778
AK094253.1 - 6 1528
AK125501.1 - 1 2159
AK127875.1 - 5 2135
AK128302.1 - 3 2811
BC001225.1 - 9 1874 3533990
BC006470.1 - 9 1830

View Full Product Details