Gene Transcripts

Gene Symbol
UQCC3
Entrez Gene ID
790955
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 3
Gene Aliases
C11orf83, CCDS41658.1, MC3DN9, UNQ655
Chromosome Location
Chr. 11: 62671654 - 62673690 on Build GRCh38
UniGene ID
Hs.569009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085372.2 NP_001078841.1 2 554
GenBank mRNA AY358935.1 - 2 554
BC090057.1 - 2 522 30347387

View Full Product Details