Gene Transcripts

Gene Symbol
Phb2
Entrez Gene ID
12034
Gene Name
prohibitin 2
Gene Aliases
AU044498, BAP, Bap37, Bcap37, REA
Chromosome Location
Chr. 6: 124712289 - 124716945 on Build GRCm38
UniGene ID
Mm.36241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007531.2 NP_031557.2 8 965
GenBank mRNA AK002334.1 - 8 931
AK011107.1 - 8 932
AK090088.1 - 8 922
AK132052.1 - 8 927
AK132058.1 - 8 954
AK135729.1 - 8 882
AK151952.1 - 8 959
AK152375.1 - 7 1614
AK153368.1 - 7 1614
AY211613.1 - 8 927
AY319418.1 - 8 817
BC145875.1 - 8 917 40131178
X78683.1 - 8 921

View Full Product Details