Gene Transcripts

Gene Symbol
Cnih2
Entrez Gene ID
12794
Gene Name
cornichon homolog 2 (Drosophila)
Gene Aliases
CNIH-2, Cnil
Chromosome Location
Chr. 19: 5092868 - 5098521 on Build GRCm38
UniGene ID
Mm.299737
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB006191.2 - 6 1116
AK141483.1 - 6 1216
BC059922.1 - 6 1202
BC103797.1 - 6 1190 30134114
CX243776.1 - 1 484

View Full Product Details