Gene Transcripts
- Gene Symbol
- Col17a1
- Entrez Gene ID
- 12821
- Gene Name
- collagen, type XVII, alpha 1
- Gene Aliases
- BP180, Bpag, Bpag2
- Chromosome Location
- Chr. 19: 47646341 - 47692042 on Build GRCm38
- UniGene ID
- Mm.1225
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC125031.1 |
- |
43, 44 |
3269 |
|
BC125032.1 |
- |
43, 44 |
3268 |
40086691 |
L08407.1 |
- |
43, 44 |
3321 |
|
View Full Product Details