Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170744.1 | NP_001164215.1 | 5 | 2335 | |
NM_009980.4 | NP_034110.1 | 7 | 1212 | ||
GenBank mRNA | AB033123.1 | - | 7 | 1077 | |
AF016508.1 | - | 5 | 684 | ||
AK134399.1 | - | 6 | 736 | ||
AK146713.1 | - | 7 | 1097 | ||
AK147871.1 | - | 7 | 1095 | ||
AK217631.1 | - | 2 | 122 | ||
BC017520.1 | - | 6 | 2180 | 4511826 |