Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145813.1 | NP_001139285.1 | 4, 5 | 444 | |
NM_010125.3 | NP_034255.2 | 4, 5 | 535 | ||
GenBank mRNA | AF049702.1 | - | 4, 5 | 526 | |
AK132484.1 | - | 4, 5 | 855 | ||
AK145214.1 | - | 4, 5 | 542 | ||
BC012424.1 | - | 4, 5 | 477 | 3584025 | |
BC021629.1 | - | 4, 5 | 488 | 2651428 |