Gene Transcripts

Gene Symbol
Esr2
Entrez Gene ID
13983
Gene Name
estrogen receptor 2 (beta)
Gene Aliases
ER[b], ERbeta, Estrb
Chromosome Location
Chr. 12: 76120419 - 76177259 on Build GRCm38
UniGene ID
Mm.2561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010157.3 NP_034287.3 7 1565
NM_207707.1 NP_997590.1 8 1619
GenBank mRNA AK054290.1 - 8 1617
AK054301.1 - 6 1427
AK054413.1 - 8 1620
AY054413.1 - 7 1080
BC141075.1 - 6 1267 9055687
BC145329.1 - 6 1267 9053885
U81451.1 - 6 1037

View Full Product Details